missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR8 Antibody (44C143), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24917APC
This item is not returnable.
View return policy
Description
TLR8 Monoclonal specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| TLR8 | |
| Monoclonal | |
| APC | |
| Q9NR97 | |
| TLR8 | |
| This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 51311 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 44C143 | |
| Flow Cytometry, Flow (Intracellular) | |
| CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human, Mouse. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction