missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR8 Antibody (935166), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8999PECY55
This item is not returnable.
View return policy
Description
TLR8 Monoclonal antibody specifically detects TLR8 in Human samples. It is validated for Flow Cytometry
Specifications
| TLR8 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 935166 | |
| Flow Cytometry | |
| CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
| Chinese hamster ovary cell line CHO-derived recombinant human TLR8, Glu27-Thr827, Accession # Q9NR97 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 51311 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction