missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR8 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-77203AF750
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TLR8 Polyclonal antibody specifically detects TLR8 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Especificaciones
| TLR8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 750 | |
| CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the middle of human TLR8. The immunogen is located within amino acids 790 - 840 of TLR8. Amino Acid Squence: CASPGDQRGKSIVSLE | |
| 0.1 mL | |
| Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 51311 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido