Learn More
TLR8 Mouse anti-Human, Clone: OTI4F3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810174
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1). The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2). Ten human homologs of TLRs (TLR1-10) have been described (3). TLR8 gene contains three exons, two of which have coding function (4). TLR8 cDNA codes for a protein of approximate molecular weight of 120 kDa (4,5).Specifications
| TLR8 | |
| Monoclonal | |
| Unconjugated | |
| TLR8 | |
| CD288 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51311 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NR97 | |
| TLR8 | |
| Human recombit protein fragment corresponding to amino acids 32-153 of human TLR8 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.