missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody (26C593.2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP2-24729AF647
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Human, Mouse, Rat, Canine, Equine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.
Specifications
| TLR9 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q9NR96 | |
| TLR9 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037). | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| 54106 | |
| Human, Mouse, Rat, Canine, Equine, Primate, Monkey | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| 26C593.2 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), KnockDown Validated | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Dog, Horse, Human, Mouse, Rat and Rhesus Monkey. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction