missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody (26C593.2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24729DL594
This item is not returnable.
View return policy
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human, Mouse, Rat, Canine, Equine, Monkey, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay, In vitro Assay, Dot Blot, Flow (Intracellular), CyTOF, KnockDown, Neutralization
Specifications
| TLR9 | |
| Monoclonal | |
| DyLight 594 | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037). | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 54106 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry | |
| 26C593.2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction