missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody (2A4C6.2E5), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-31150R
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TLR9 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TLR9 | |
| Partial recombinant protein made to an internal portion of mouse TLR9 (between amino acids 500-800) [UniProt Q9EQU3] | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| Mouse | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2A4C6.2E5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction