missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody (980914), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36582R
This item is not returnable.
View return policy
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TLR9 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 980914 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Human TLR9 synthetic peptide, Accession # Q9NR96 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 54106 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction