Learn More
TLR9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-77254-0.025mg
 View more versions of this product
Product Code. 18722934
  • € 178.00 / 0.03mg
Description
TLR9 Polyclonal specifically detects TLR9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLR9
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
CD289, CD289 antigen, toll-like receptor 9
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL
AAH32713
TLR9
Antibody was raised against a peptide corresponding to 16 amino acids near the carboxy terminus of human TLR9. The immunogen is located within amino acids 960 - 1010 of TLR9. Amino Acid Squence: LSPDGRRSRYVRLRQR
0.025 mg
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
54106
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.