missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR9 Monoclonal Antibody (5G5), Biotin
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549118
This item is not returnable.
View return policy
Description
The monoclonal antibody 5G5 reacts with RAW macrophages and TLR9 transfected HEK293 cells, and is cross reactive with canine TLR9.
The protein encoded by this gene is a member of the Toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is preferentially expressed in immune cell rich tissues, such as spleen, lymph node, bone marrow and peripheral blood leukocytes. Studies in mice and human indicate that this receptor mediates cellular response to unmethylated CpG dinucleotides in bacterial DNA to mount an innate immune response.
Specifications
| TLR9 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Q9NR96 | |
| Tlr9 | |
| Purified fusion protein of extracellular domain of human TLR9 (AA 1-815) and human IgGFc. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5G5 | |
| Biotin | |
| Tlr9 | |
| CD289; Tlr9; toll like receptor 9; toll-like receptor 9; UNQ5798/PRO19605 | |
| Mouse | |
| Protein G | |
| RUO | |
| 54106 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction