missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9, Mouse anti-Human, Clone: 2C3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00054106-M14.100ug
This item is not returnable.
View return policy
Description
Sequence: DMHGIFFRSLDETTLRPLARLPMLQTLRLQMNFINQAQLGIFRAFPGLRYVDLSDNRISGASELTATMGEADGGEKVWLQPGDLAPAPVDTPSSEDFRPNCSTLSpecifications
| TLR9 | |
| Monoclonal | |
| Unconjugated | |
| toll-like receptor 9 | |
| CD289 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2C3 | |
| In 1x PBS, pH 7.4 | |
| NM_017442 | |
| TLR9 | |
| TLR9 (NP_059138, 370 a.a. ∼ 473 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 54106 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction