missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLX/NR2E1 Antibody (SR1840), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22225-50ul
This item is not returnable.
View return policy
Description
TLX/NR2E1 Monoclonal antibody specifically detects TLX/NR2E1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TLX/NR2E1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1840 | |
| Western Blot 1:500-1:2000 | |
| hTll, nuclear receptor subfamily 2, group E, member 1, Nuclear receptor TLX, Protein tailless homolog, tailes-related receptor, tailless homolog, Tll, TLXnuclear receptor subfamily 2 group E member 1, XTLL | |
| A synthesized peptide derived from human TLX/NR2E1 (Uniprot #: Q9Y466) | |
| 50 μg | |
| Neuroscience | |
| 7101 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction