missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA124538
This item is not returnable.
View return policy
Description
PA1-24538 detects TLX in human samples.
Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
Specifications
| TLX | |
| Polyclonal | |
| Unconjugated | |
| NR2E1 | |
| fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100072159, 100155153, 100328698, 475017, 528156, 7101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y466 | |
| NR2E1 | |
| Synthetic 15 amino acid peptide from a N-terminal region of human NR2E1. | |
| 25 μg | |
| Primary | |
| Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction