Learn More
Invitrogen™ TLX3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA534555
Product Code. 15325481
  • € 578.00 / 100µg
Description
A suggested positive control is human spleen tissue lysate. PA5-34555 can be used with blocking peptide PEP-1598.
The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX3, also known as homeobox11-like1 (HOX11L2), is a DNA-binding nuclear transcription factor that is expressed in a subset of the primary sensory nervous system. TLX3, along with the related TLX1, is a selector transcription factor that promotes an excitatory glutamatergic neuronal phenotype over an inhibitory GABAergic phenotype opposing LBX1 signals during dorsal spinal cord development. Chromosomal translocations of the TLX3 gene have been shown to result in some forms of T-cell acute lymphoblastic leukemia (T-ALL).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLX3
Polyclonal
Unconjugated
TLX3
homeo box 11-like 2; homeobox 11-like 2; homeobox protein Hox-11L2; Homeobox TLX-3; HOX11L2; Respiratory neuron homeobox protein; RGD1564190; RNX; T cell leukemia, homeobox 3; T-cell leukemia homeobox 3; T-cell leukemia homeobox protein 3; T-cell leukemia, homeobox 3; Tlx1l2; TLX3
Rabbit
Antigen affinity chromatography
RUO
27140, 30012, 497881
4°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O43711, O55144
TLX3
A 16 amino acid peptide near the amino terminus of human TLX3.
100 μg
Primary
Human, Mouse, Rat