missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM2D2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98504PCP
This item is not returnable.
View return policy
Description
TM2D2 Polyclonal antibody specifically detects TM2D2 in Human samples. It is validated for Western Blot
Specifications
| TM2D2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83877 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| PerCP | |
| BBP-like protein 1, Beta-amyloid-binding protein-like protein 1, BLP1MGC125814, MGC125813, TM2 domain containing 2, TM2 domain-containing protein 2 | |
| Produced in rabbits immunized with E. coli-derived Human TM2D2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction