missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TM2D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561448
This item is not returnable.
View return policy
Description
Immunogen sequence: TAEPELTSAG AAQPEGPGGA ASWEYGDPHS PVILCSYLPD EFIECEDPVD HVGNAT Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. This gene has multiple alternatively spliced transcript variants which encode two different isoforms.
Specifications
| TM2D2 | |
| Polyclonal | |
| Unconjugated | |
| TM2D2 | |
| 2410018G23Rik; BBP-like protein 1; Beta-amyloid-binding protein-like protein 1; BLP1; RGD1306769; si:dkey-49o11.3; TM2 domain containing 2; TM2 domain containing 2, isoform a; TM2 domain-containing protein 2; TM2 domain-containing protein 2-like; TM2 domain-containing protein 2-like protein; TM2D2; Unknown (protein for MGC:133974); zgc:92201 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BX73 | |
| TM2D2 | |
| Recombinant protein corresponding to Human TM2D2. Recombinant protein control fragment (Product #RP-99887). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction