Learn More
TM4SF1/L6 Antibody (877621) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8164M
Product Code. 18610370
  • ONLINE EXCLUSIVE PRICE
    € 695.00
    € 657.30 / 0.10mL
    Save € 37.70
    5% Off
Description
TM4SF1/L6 Monoclonal antibody specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TM4SF1/L6
Monoclonal
DyLight 594
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
877621
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
L6, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1
NS0 mouse myeloma cell line transfected with human TM4SF1/L6, Met1-Cys202, Accession # P30408
0.1 mL
Cancer, Cell Cycle and Replication
4071
Store at 4°C in the dark.