missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM4SF1/L6 Antibody (877621), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8164MFV610
This item is not returnable.
View return policy
Description
TM4SF1/L6 Monoclonal antibody specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Specifications
| TM4SF1/L6 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 4071 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 877621 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
| L6, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1 | |
| NS0 mouse myeloma cell line transfected with human TM4SF1/L6, Met1-Cys202, Accession # P30408 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction