missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM4SF18 Antibody (211116), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3509PECY55
This item is not returnable.
View return policy
Description
TM4SF18 Monoclonal antibody specifically detects TM4SF18 in Human samples. It is validated for Flow Cytometry
Specifications
| TM4SF18 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 116441 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 211116 | |
| Flow Cytometry | |
| L6D | |
| NS0 mouse myeloma cell line transfected with human TM4SF18, Met1-Ile201, Accession # Q96CE8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction