missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM4SF4 Antibody (832441) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7998M
This item is not returnable.
View return policy
Description
TM4SF4 Monoclonal antibody specifically detects TM4SF4 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TM4SF4 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 832441 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ31015, il-TMP, ILTMPintestinal and liver (il) tetraspan membrane protein, Intestine and liver tetraspan membrane protein, transmembrane 4 L six family member 4, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4 | |
| NS0 mouse myeloma cell line transfected with human TM4SF4, Accession # P48230 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 7104 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction