missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TM4SF5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17988842
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988842 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988842 Supplier Invitrogen™ Supplier No. PA5144107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TM4SF5 Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TM4SF5
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Tm4sf5
Gene Accession No. O14894
Gene Alias 2010003F10Rik; AI323987; Tetraspan transmembrane protein L6H; TM4SF5; transmembrane 4 L six family member 5; transmembrane 4 L6 family member 5; transmembrane 4 superfamily member 5
Gene Symbols Tm4sf5
Host Species Rabbit
Immunogen Recombinant Human Transmembrane 4 L6 family member 5 protein (112-157AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9032
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.