missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM9SF2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98514MFV610
This item is not returnable.
View return policy
Description
TM9SF2 Polyclonal antibody specifically detects TM9SF2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TM9SF2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9375 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| FLJ26287, MGC117391, P76, transmembrane 9 superfamily member 2, transmembrane protein 9 superfamily member 2,76 kDa membrane protein | |
| Produced in rabbits immunized with E. coli-derived Human TM9SF2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction