missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15596092
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15596092 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15596092 Supplier Invitrogen™ Supplier No. PA543535

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: YCWCWDLEAG FPSYAEFDIS GNVLGLIFNQ GMIWMGSFYA PGLVGINVLR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%; Zebrafish: 100%.

This gene is considered a member of a gene family predicted to encode transmembrane proteins. The specific function of this gene is unknown; however, expression in the inner ear suggests that it may be crucial for normal auditory function. Mutations in this gene may underlie hereditary disorders of balance and hearing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMC2
Gene Accession No. Q8TDI7
Gene Alias 4933416G09Rik; Beethoven; Bth; C20orf145; CWEA1; CWEA2; dJ686C3.3; dn; TMC2; transmembrane channel like 2; transmembrane channel-like 2; transmembrane channel-like gene family 2; transmembrane channel-like protein 2; transmembrane cochlear-expressed gene 2; transmembrane cochlear-expressed protein 2; transmembrane, cochlear expressed 2; UNQ907/PRO1928
Gene Symbols TMC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TMC2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117532
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.