missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMCC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569174
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
TMCC2 may be involved in the regulation of the proteolytic processing of the amyloid precursor protein (APP) possibly also implicating APOE.
Specifications
| TMCC2 | |
| Polyclonal | |
| Unconjugated | |
| TMCC2 | |
| 1110063G11Rik; cerebral protein 11; HUCEP11; hucep-11; Kiaa0481; RGD1311960; TMCC2; transmembrane and coiled-coil domain family 2; transmembrane and coiled-coil domains 2; transmembrane and coiled-coil domains protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9911 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75069 | |
| TMCC2 | |
| synthetic peptide directed towards the N terminal region of human TMCC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction