missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMCC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30019592
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019592 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019592 Supplier Invitrogen™ Supplier No. PA5148330

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMCC2 Polyclonal Antibody for IHC (P)

TMCC2 may be involved in the regulation of the proteolytic processing of the amyloid precursor protein (APP) possibly also implicating APOE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene TMCC2
Gene Accession No. O75069
Gene Alias 1110063G11Rik; cerebral protein 11; HUCEP11; hucep-11; Kiaa0481; RGD1311960; TMCC2; transmembrane and coiled-coil domain family 2; transmembrane and coiled-coil domains 2; transmembrane and coiled-coil domains protein 2
Gene Symbols TMCC2
Host Species Rabbit
Immunogen E. coli-derived Human TMCC2 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.