missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMCO4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15937734
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15937734 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15937734 Supplier Invitrogen™ Supplier No. PA564955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QWLELSEAVLP TMTAFASGLG GEGADVFVQI LLKDPILKDD PTVITQDLLS FSLKDGHYDA RARVLVCHMT SLLQVPLEEL DVLEEMFLES LKEIKEEESE MAEASRKKKE Highest antigen sequence identity to the following orthologs - mouse 78%, rat 80%.

TMCO4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCO4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMCO4
Gene Accession No. Q5TGY1
Gene Alias TMCO4; Transmembrane and coiled-coil domain-containing protein 4; transmembrane and coiled-coil domains 4
Gene Symbols TMCO4
Host Species Rabbit
Immunogen Recombinant protein fragment of Human TMCO4 (Product #RP-91007)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255104
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.