missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED1 Antibody (1009524), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB22431D
This item is not returnable.
View return policy
Description
TMED1 Monoclonal antibody specifically detects TMED1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| TMED1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11018 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 1009524 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| Il1rl1l, IL1RL1LGIL1RL1-binding protein, interleukin 1 receptor-like 1 ligand, Interleukin-1 receptor-like 1 ligand, MGC1270, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1 | |
| Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction