missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED1 Antibody (1009524) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB22431JF525
This item is not returnable.
View return policy
Description
TMED1 Monoclonal antibody specifically detects TMED1 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| TMED1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Il1rl1l, IL1RL1LGIL1RL1-binding protein, interleukin 1 receptor-like 1 ligand, Interleukin-1 receptor-like 1 ligand, MGC1270, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1 | |
| Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 1009524 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 11018 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction