missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMED5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17949712
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949712 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949712 Supplier Invitrogen™ Supplier No. PA5144635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive control: Rat liver tissue lysate, A431 cell lysate, Jurkat cell lysate, Hela cell lysate, SiHa cell lysate, rat bladder tissue, mouse large intestine tissue. Predicted band size: 26 kDa Subcellular Location: Cis-Golgi network membrane, endoplasmic reticulum membrane, endoplasmic reticulum-Golgi intermediate compartment membrane.

The TMED5 gene encodes the transmembrane emp24 domain-containing protein 5, a member of the TMED family, which is involved in intracellular protein trafficking. This family is essential for the transport of proteins between the endoplasmic reticulum and the Golgi apparatus. TMED5 is a member of the γ subfamily and has been shown to regulate vital cellular processes, including protein secretion and signal transduction. Dysregulation of TMED5 has been implicated in various diseases, including hepatocellular carcinoma (HCC). Increased expression of TMED5 mRNA in HCC tissues is associated with poorer overall survival rates compared to normal tissues, suggesting its role in tumor progression and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TMED5
Gene Accession No. Q6AXN3, Q9CXE7, Q9Y3A6
Gene Alias CGI-100; p24 family protein gamma-2; p24g2; p24gamma2; p28; TMED5; Transmembrane emp24 domain-containing protein 5; transmembrane emp24 protein transport domain containing 5; transmembrane p24 trafficking protein 5; UNQ397/PRO733
Gene Symbols TMED5
Host Species Rabbit
Immunogen Recombinant protein within Human TMED5 aa 1-200/229.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289883, 50999, 73130
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.