Learn More
Novus Biologicals™ TMEFF1/Tomoregulin-1 Antibody (Bluefin patent anti-TMEFF1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28556-50ug
 View more versions of this product
Product Code. 30044156
  • € 213.00 / 50µg
Description
TMEFF1/Tomoregulin-1 Monoclonal antibody specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEFF1/Tomoregulin-1
Monoclonal
Unconjugated
Q8IYR6
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Bluefin patent anti-TMEFF1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
C9orf2, H7365member 1, tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1
TMEFF1/ Tomoregulin-1
50 μg
Core ESC Like Genes, Stem Cell Markers
8577