missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEFF2 Monoclonal Antibody (OTI3D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504464S
This item is not returnable.
View return policy
Description
TMEFF2 Monoclonal antibody specifically detects TMEFF2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| TMEFF2 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q06203 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5471 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| OTI3D1 | |
| Unconjugated | |
| Ppat | |
| 4832418D20Rik; 7630402F16Rik; cancer/testis antigen family 120, member 2; CT120.2; HPP1; Hyperplastic polyposis protein 1; MNCb-1026; TENB2; TMEFF2; tomoregulin; tomoregulin-2; TPEF; TR; TR-2; transmembrane protein TENB2; transmembrane protein with EGF like and two follistatin like domains 2; transmembrane protein with EGF-like and two follistatin-like domains; transmembrane protein with EGF-like and two follistatin-like domains 2; UNQ178/PRO204 | |
| Human recombinant protein fragment corresponding to amino acids 42-278 of human PPAT produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction