missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEFF2 Monoclonal Antibody (OTI4G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504465
This item is not returnable.
View return policy
Description
TMEFF2 may be a survival factor for hippocampal and mesencephalic neurons. The shedded form up-regulates cancer cell proliferation, probably by promoting ERK1/2 phosphorylation.Specifications
| TMEFF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UIK5 | |
| TMEFF2 | |
| Full length human recombit protein of human TMEFF2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| OTI4G2 | |
| Unconjugated | |
| TMEFF2 | |
| CT120.2, HPP1, TENB2, TPEF, TR, TR-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23671 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction