missing translation for 'onlineSavingsMsg'
Learn More

TMEFF2/Tomoregulin-2 Antibody (Janssen patent anti-TMEFF2), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 30116181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30116181 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30116181 Supplier Novus Biologicals Supplier No. NBP327983AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEFF2/Tomoregulin-2
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Janssen patent anti-TMEFF2
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2
Host Species Human
Immunogen TMEFF2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokines and Growth Factors
Primary or Secondary Primary
Gene ID (Entrez) 23671
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.