Learn More
Novus Biologicals™ TMEFF2/Tomoregulin-2 Antibody (Janssen patent anti-TMEFF2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27983-100ug
 View more versions of this product
Product Code. 30047767
  • € 338.00 / 100µg
Description
TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEFF2/Tomoregulin-2
Monoclonal
Unconjugated
Q9UIK5
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Janssen patent anti-TMEFF2
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2
TMEFF2
100 μg
Cancer, Cell Biology, Cytokines and Growth Factors
23671