missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM100 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 232.00 - € 483.00
Specifications
| Antigen | TMEM100 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673342
|
Novus Biologicals
NBP2-94746-0.02ml |
0.02 mL |
€ 232.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633742
|
Novus Biologicals
NBP2-94746-0.1ml |
0.1 mL |
€ 483.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM100 Polyclonal antibody specifically detects TMEM100 in Human, Mouse samples. It is validated for Western BlotSpecifications
| TMEM100 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FLJ10970, FLJ37856, transmembrane protein 100 | |
| A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human TMEM100 (NP_001093110.1). ELSCYRCIIPFAVVVFIAGIVVTAVAYSFNSHGSIISIFGLVVLSSGLFLLASSALCWKVRQRSKKAKRRESQTALVANQRSLFA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55273 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title