Learn More
TMEM100 Mouse anti-Canine, Human, Clone: OTI1F8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500508
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Plays a role during embryonic arterial endothelium differentiation and vascular morphogenesis through the ACVRL1 receptor-dependent signaling pathway upon stimulation by bone morphogenetic proteins, such as GDF2/BMP9 and BMP10.Specifications
| TMEM100 | |
| Monoclonal | |
| Unconjugated | |
| TMEM100 | |
| transmembrane protein 100 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55273, 609661 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NV29 | |
| TMEM100 | |
| Full-length protein expressed in 293T cell transfected with human TMEM100 expression vector | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.