missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM100 Mouse anti-Human, Clone: OTI2D3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500499
This item is not returnable.
View return policy
Description
Plays a role during embryonic arterial endothelium differentiation and vascular morphogenesis through the ACVRL1 receptor-dependent signaling pathway upon stimulation by bone morphogenetic proteins, such as GDF2/BMP9 and BMP10.Specifications
| TMEM100 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NV29 | |
| TMEM100 | |
| Full-length protein expressed in 293T cell transfected with human TMEM100 expression vector | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D3 | |
| Unconjugated | |
| TMEM100 | |
| transmembrane protein 100 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55273 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction