Learn More
Invitrogen™ TMEM102 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570151
Product Code. 15818563
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%.
Selectively involved in CSF2 deprivation-induced apoptosis via a mitochondria-dependent pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEM102
Polyclonal
Unconjugated
TMEM102
4632403M07Rik; Cbap; common beta-chain associated protein; common beta-chain-associated protein; LOW QUALITY PROTEIN: transmembrane protein 102; RGD1559450; TMEM102; Tmem102-ps; transmembrane protein 102; transmembrane protein 102, pseudogene
Rabbit
Affinity chromatography
RUO
380705
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q3UPR7
TMEM102
synthetic peptide corresponding to a region of mouse TMEM102 (aa 64-113).