missing translation for 'onlineSavingsMsg'
Learn More

TMEM102 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16834075 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16834075 150 μL 150µL
16844075 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16834075 Supplier Proteintech Supplier No. 255051AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Selectively involved in CSF2 deprivation-induced apoptosis via a mitochondria-dependent pathway.

Specifications

Antigen TMEM102
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TMEM102
Gene Accession No. Q3UPR7, Q8N9M5
Gene Alias CBAP, TMEM102, transmembrane protein 102
Gene Symbols Tmem102
Host Species Rabbit
Immunogen TMEM102 Fusion Protein Ag22213
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284114, 380705
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.