missing translation for 'onlineSavingsMsg'
Learn More

TMEM103 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18580673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18580673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18580673 Supplier Novus Biologicals Supplier No. NBP298169G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM103 Polyclonal antibody specifically detects TMEM103 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM103
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias Angiotonin-transactivated protein 1, ATP1, C3orf75, chromosome 3 open reading frame 75, elongator acetyltransferase complex subunit 6, ELP6, FLJ20211, Protein TMEM103, TMEM103, transmembrane protein 103
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TMEM103 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54859
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.