missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM103 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55872
This item is not returnable.
View return policy
Description
TMEM103 Polyclonal specifically detects TMEM103 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| C3orf75/ELP6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Angiotonin-transactivated protein 1, ATP1, C3orf75, chromosome 3 open reading frame 75, elongator acetyltransferase complex subunit 6, ELP6, FLJ20211, Protein TMEM103, TMEM103, transmembrane protein 103 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54859 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ELP6 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LGVSLTMARERGQLVFLEGLKSAVDVVFQAQKEPHPLQFLREANAGNLKPLFEFVREALKPVDSGEAR | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only
Spot an opportunity for improvement?