missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM106B Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82027JF549
This item is not returnable.
View return policy
Description
TMEM106B Polyclonal antibody specifically detects TMEM106B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TMEM106B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| transmembrane protein 106B | |
| TMEM106B antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human TMEM106B. The immunogen is located within the first 50 amino acids of TMEM106B. Amino Acid Squence: NSEVHNEDGRNGDVS | |
| 0.1 mL | |
| Neuroscience | |
| 54664 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction