missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM107 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572774
This item is not returnable.
View return policy
Description
TMEM107 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Gene ontology (GO) annotation includes embryonic digit morphogenesis; neural tube patterning; non-motile cilium assembly; protein localization to ciliary transition zone.
Specifications
| TMEM107 | |
| Polyclonal | |
| Unconjugated | |
| TMEM107 | |
| 1110004B13Rik; 2810049P21Rik; DC20; GRVS638; PRO1268; TMEM107; Transmembrane protein 107; UNQ638/PRO1268 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6UX40 | |
| TMEM107 | |
| TMEM107 antibody for a 19 amino acid peptide near the amino terminus of human TMEM107. The immunogen is within amino acids 20 - 70 of TMEM107. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction