missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM108 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21447MFV500
This item is not returnable.
View return policy
Description
TMEM108 Polyclonal antibody specifically detects TMEM108 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| TMEM108 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| cancer/testis antigen 124, CT124, KIAA1690, MGC3040, transmembrane protein 108 | |
| Produced in rabbits immunized with E. coli-derived Human TMEM108 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 66000 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction