Learn More
Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2), Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech 65122-1-IG-100UG
 View more versions of this product
Product Code. 30236831
  • € 44.00 / 100µg
Description
The MHC Class II (I-A/I-E) antibody from Proteintech is a rat monoclonal antibody generated with activated c57bl/6 mouse spleen cells of mouse MHC Class II (I-A/I-E). This antibody recognizes mouse antigen. The MHC Class II (I-A/I-E) antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
MHC Class II (I-A/I-E)
Monoclonal
0.5 mg/mL
Rat
Affinity purification
RUO
14960
Store at 2-8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
M5/114.15.2
Unconjugated
Activated C57BL/6 mouse spleen cells
100 μg
Primary
Mouse
Flow Antibody
IgG2b κ