Learn More
Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65122-1-IG-100UG
This item is not returnable.
View return policy
Description
The MHC Class II (I-A/I-E) antibody from Proteintech is a rat monoclonal antibody generated with activated c57bl/6 mouse spleen cells of mouse MHC Class II (I-A/I-E). This antibody recognizes mouse antigen. The MHC Class II (I-A/I-E) antibody has been validated for the following applications: FC analysis.Specifications
| MHC Class II (I-A/I-E) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Affinity purification | |
| RUO | |
| 14960 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| M5/114.15.2 | |
| Unconjugated | |
| Activated C57BL/6 mouse spleen cells | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2b κ |