missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM111 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | TMEM111 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236830
|
Proteintech
24645-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235407
|
Proteintech
24645-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TMEM111 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TMEM111. This antibody recognizes human, mouse, rat antigen. The TMEM111 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| TMEM111 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| POB | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 55831 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.