missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM119 Antibody (2699A) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB103131JF525
This item is not returnable.
View return policy
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry
Specifications
| TMEM119 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| OBIF, Osteoblast Induction Factor, TMEM119 | |
| Chinese Hamster Ovary cell line CHO derived human TREM119, Arg26-Met96, Accession # Q4V9L6.1 | |
| RUO | |
| 338773 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunohistochemistry | |
| 2699A | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction