missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM131 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21369H
This item is not returnable.
View return policy
Description
TMEM131 Polyclonal antibody specifically detects TMEM131 in Human samples. It is validated for Western Blot
Specifications
| TMEM131 | |
| Polyclonal | |
| Western Blot | |
| KIAA02572610524E03Rik, PRO1048, Protein RW1, RW1CC28, transmembrane protein 131, YR-23 | |
| The immunogen recognized by this antibody maps to a region between residue 1779 and 1829 of human transmembrane protein 131 using the numbering given in entry BAA13387.1 (GeneID 23505). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23505 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction