missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM158 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549532
This item is not returnable.
View return policy
Description
Predicted to react with bovine, mouse and rat based on sequence homology.
Receptor for brain injury-derived neurotrophic peptide (BINP), a synthetic 13-mer peptide.
Specifications
| TMEM158 | |
| Polyclonal | |
| Unconjugated | |
| TMEM158 | |
| 2310037P21Rik; 40 kDa BINP-binding protein; BBP; BINP binding protein p40BBP; BINP receptor; brain injury-derived neurotrophic peptide (BINP) binding protein; brain specific binding protein; brain-specific binding protein; HBBP; Mbbp; p40BBP; Ras induced senescence 1; Ras-induced senescence 1; ras-induced senescence protein 1; Ris1; Tmem158; Transmembrane protein 158; transmembrane protein 158 (gene/pseudogene) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 25907 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8WZ71 | |
| TMEM158 | |
| KLH conjugated synthetic peptide between 186-218 amino acids from human TMEM158. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction