missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM163 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114329
This item is not returnable.
View return policy
Description
TMEM163 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
May bind zinc and other divalent cations and recruit them to vesicular organelles. [UniProt].
Specifications
| TMEM163 | |
| Polyclonal | |
| Unconjugated | |
| TMEM163 | |
| 2610024A01Rik; DC29; RGD1306212; Sv31; synaptic vesicle membrane protein of 31 kDa; synaptic vesicle protein of 30 kD; Tmem163; Transmembrane protein 163 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360839, 72160, 81615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| A9CMA6, Q8C996, Q8TC26 | |
| TMEM163 | |
| Synthetic peptide within Human TMEM163 aa 55-104. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction